siRNA Details
Gene Symbol: TMEM72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123376.2 | NP_001116848.1 | 4 | 439 | ||
XM_011540069.2 | XP_011538371.1 | 4 | 3241 | |||
XM_011540070.2 | XP_011538372.1 | 4 | 3187 | |||
XM_011540071.2 | XP_011538373.1 | 5 | 548 | |||
GenBank mRNA | AB235418.1 | - | 4 | 394 | ||
BC127789.1 | - | 3 | 372 | 40133552 | ||
BC127790.1 | - | 4 | 439 | 40133554 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
