siRNA Details
Gene Symbol: LRRC66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024611.2 | NP_001019782.1 | 2 | 578 | ||
XM_011534383.2 | XP_011532685.1 | 3 | 2299 | |||
GenBank mRNA | CR749811.1 | - | 2 | 578 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
