siRNA Details
Gene Symbol: PLAC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012973.1 | NP_001012991.1 | 3 | 438 | ||
GenBank mRNA | BC066348.1 | - | 3 | 438 | 30339526 | |
BC090922.1 | - | 3 | 438 | 30331098 | ||
BC117332.1 | - | 3 | 266 | 40125883 | ||
BC117334.1 | - | 3 | 266 | 40125885 |
Gene Symbol: LOC389033 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026740.1 | 2 | 154 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
