siRNA Details
Gene Symbol: FAM159A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042693.2 | NP_001036158.1 | 3 | 536 | ||
XM_011541377.2 | XP_011539679.1 | 3 | 1805 | |||
XM_011541378.2 | XP_011539680.1 | 4 | 1981 | |||
GenBank mRNA | AY358621.1 | - | 3 | 310 | ||
BG192125.1 | - | 2 | 208 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
