siRNA Details
Gene Symbol: CISD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136498.1 | NP_001129970.1 | 4 | 349 | ||
GenBank mRNA | AK097047.1 | - | 3 | 600 | ||
AK098433.1 | - | 2 | 130 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
