siRNA Details
Gene Symbol: C6orf222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010903.4 | NP_001010903.3 | 11 | 2062 | ||
XM_011514596.2 | XP_011512898.1 | 11 | 2915 | |||
XM_011514597.2 | XP_011512899.1 | 11 | 2189 | |||
XM_011514598.2 | XP_011512900.1 | 10 | 1572 | |||
GenBank mRNA | BC140873.1 | - | 11 | 1985 | 9021741 | |
BX648558.1 | - | 11 | 2063 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
