siRNA Details
Gene Symbol: C17orf100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105520.1 | NP_001098990.1 | 1 | 629 | ||
GenBank mRNA | BC038956.1 | - | 1 | 650 | ||
BC082254.1 | - | 1 | 629 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
