siRNA Details
Gene Symbol: C19orf67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277378.1 | NP_001264307.1 | 4, 5 | 944 | ||
XM_011528184.2 | XP_011526486.1 | 4, 5 | 1299 | |||
GenBank mRNA | DN831393.1 | - | 2, 3 | 298 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
