siRNA Details
Gene Symbol: KIAA1109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015312.3 | NP_056127.2 | 5 | 579 | ||
XM_005263282.1 | XP_005263339.1 | 7 | 907 | |||
XM_005263287.1 | XP_005263344.1 | 7 | 907 | |||
XM_006714344.1 | XP_006714407.1 | 7 | 907 | |||
XM_011532319.1 | XP_011530621.1 | 7 | 907 | |||
XM_011532320.2 | XP_011530622.1 | 8 | 1002 | |||
XM_011532321.2 | XP_011530623.1 | 6 | 709 | |||
XM_011532322.1 | XP_011530624.1 | 7 | 907 | |||
XM_011532323.1 | XP_011530625.1 | 7 | 907 | |||
XM_011532324.1 | XP_011530626.1 | 7 | 907 | |||
XM_011532325.1 | XP_011530627.1 | 7 | 907 | |||
XM_011532326.1 | XP_011530628.1 | 7 | 907 | |||
XM_011532327.1 | XP_011530629.1 | 8 | 953 | |||
XM_017008695.1 | XP_016864184.1 | 7 | 907 | |||
XM_017008696.1 | XP_016864185.1 | 7 | 907 | |||
XM_017008697.1 | XP_016864186.1 | 7 | 907 | |||
XM_017008699.1 | XP_016864188.1 | 7 | 907 | |||
GenBank mRNA | AK124902.1 | - | 1 | 31 | ||
DQ335469.1 | - | 5 | 579 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
