siRNA Details
Gene Symbol: FAM25C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137548.2 | NP_001131020.1 | 3 | 333 | ||
GenBank mRNA | BX093303.1 | - | 3 | 388 |
Gene Symbol: FAM25A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146157.2 | NP_001139629.1 | 3 | 343 |
Gene Symbol: FAM25BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104039.1 | 3 | 333 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
