siRNA Details
Gene Symbol: PLEKHM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080475.2 | NP_001073944.1 | 8 | 9565 | ||
XM_005246545.3 | XP_005246602.1 | 7 | 9407 | |||
XM_017004072.1 | XP_016859561.1 | 7 | 9750 | |||
XM_017004073.1 | XP_016859562.1 | 7 | 8629 | |||
GenBank mRNA | AF462444.1 | - | 2 | 1027 | ||
BC042862.1 | - | 1 | 1240 | |||
BX538248.1 | - | 1 | 1601 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
