siRNA Details
Gene Symbol: PLEKHM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080475.2 | NP_001073944.1 | 8 | 8747 | ||
XM_005246545.3 | XP_005246602.1 | 7 | 8589 | |||
XM_011511160.2 | XP_011509462.1 | 9 | 9082 | |||
XM_017004072.1 | XP_016859561.1 | 7 | 8932 | |||
XM_017004073.1 | XP_016859562.1 | 7 | 7811 | |||
GenBank mRNA | AF462444.1 | - | 1 | 427 | ||
BC042862.1 | - | 1 | 422 | |||
BX538248.1 | - | 1 | 783 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
