siRNA Details
Gene Symbol: RNF222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146684.2 | NP_001140156.1 | 3 | 505 | ||
XM_011523978.2 | XP_011522280.1 | 3 | 482 | |||
XM_011523979.2 | XP_011522281.1 | 3 | 590 | |||
XM_011523980.2 | XP_011522282.1 | 2 | 374 | |||
XM_011523981.2 | XP_011522283.1 | 2 | 318 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
