siRNA Details
Gene Symbol: NHLRC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012754.3 | NP_001012772.1 | 7 | 1908 | ||
NM_001017370.2 | NP_001017370.1 | 6 | 1707 | |||
NR_073109.1 | 7 | 1650 | ||||
GenBank mRNA | BC052492.1 | - | 7 | 1588 | ||
BC140877.1 | - | 7 | 1834 | 9021745 | ||
BC140878.1 | - | 7 | 1834 | 9021746 | ||
BX647872.1 | - | 6 | 1597 | |||
BX648122.1 | - | 7 | 1858 | |||
CR749318.1 | - | 6 | 1629 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
