siRNA Details
Gene Symbol: NCKAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207363.2 | NP_997246.2 | 20 | 6128 | ||
NM_207481.3 | NP_997364.3 | 18 | 2171 | |||
XM_005263659.3 | XP_005263716.1 | 18 | 6577 | |||
XM_005263660.3 | XP_005263717.1 | 18 | 6092 | |||
XM_011511097.2 | XP_011509399.1 | 20 | 6765 | |||
XM_011511098.2 | XP_011509400.1 | 20 | 6762 | |||
XM_011511099.2 | XP_011509401.1 | 19 | 6160 | |||
XM_011511100.2 | XP_011509402.1 | 19 | 6258 | |||
XM_011511101.1 | XP_011509403.1 | 17 | 5668 | |||
XM_011511102.2 | XP_011509404.1 | 18 | 10094 | |||
XM_011511103.2 | XP_011509405.1 | 16 | 5896 | |||
XM_011511104.1 | XP_011509406.1 | 16 | 5547 | |||
XM_011511105.2 | XP_011509407.1 | 16 | 6338 | |||
XM_017003974.1 | XP_016859463.1 | 19 | 5888 | |||
XM_017003975.1 | XP_016859464.1 | 18 | 6190 | |||
XM_017003976.1 | XP_016859465.1 | 20 | 6786 | |||
XM_017003977.1 | XP_016859466.1 | 17 | 6696 | |||
XM_017003978.1 | XP_016859467.1 | 13 | 6106 | |||
XM_017003979.1 | XP_016859468.1 | 17 | 2678 | |||
XM_017003980.1 | XP_016859469.1 | 15 | 2072 | |||
GenBank mRNA | AK124659.1 | - | 7 | 1000 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
