siRNA Details
Gene Symbol: INSC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031853.4 | NP_001027024.3 | 9 | 1290 | ||
NM_001042536.2 | NP_001036001.1 | 9 | 1214 | |||
NM_001278313.1 | NP_001265242.1 | 8 | 1176 | |||
NM_001278314.1 | NP_001265243.1 | 9 | 1281 | |||
NM_001278315.1 | NP_001265244.1 | 8 | 1176 | |||
NM_001278316.1 | NP_001265245.1 | 7 | 1050 | |||
XM_006718227.3 | XP_006718290.1 | 10 | 1343 | |||
XM_011520085.1 | XP_011518387.1 | 9 | 1218 | |||
XM_011520086.1 | XP_011518388.1 | 9 | 1218 | |||
XM_017017697.1 | XP_016873186.1 | 10 | 1255 | |||
XM_017017698.1 | XP_016873187.1 | 9 | 1218 | |||
GenBank mRNA | AB231745.1 | - | 8 | 1170 | ||
AB231746.1 | - | 9 | 1275 | |||
AB231747.1 | - | 7 | 1044 | |||
AB236158.1 | - | 9 | 1214 | |||
AB236159.1 | - | 9 | 1290 | |||
BC127700.1 | - | 8 | 1176 | 40127962 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
