siRNA Details
Gene Symbol: KIAA1257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011513050.2 | XP_011511352.2 | 10 | 1777 | ||
XM_017006939.1 | XP_016862428.1 | 11 | 2332 | |||
XM_017006940.1 | XP_016862429.1 | 11 | 2328 | |||
XM_017006941.1 | XP_016862430.1 | 11 | 1905 | |||
XM_017006942.1 | XP_016862431.1 | 11 | 2299 | |||
XM_017006943.1 | XP_016862432.1 | 11 | 2057 | |||
XM_017006944.1 | XP_016862433.1 | 10 | 1877 | |||
XM_017006945.1 | XP_016862434.1 | 10 | 1681 | |||
XM_017006946.1 | XP_016862435.1 | 9 | 1488 | |||
XM_017006947.1 | XP_016862436.1 | 8 | 1372 | |||
XM_017006948.1 | XP_016862437.1 | 10 | 1549 | |||
XM_017006949.1 | XP_016862438.1 | 9 | 1496 | |||
XM_017006950.1 | XP_016862439.1 | 9 | 6983 | |||
XM_017006951.1 | XP_016862440.1 | 9 | 7088 | |||
XM_017006952.1 | XP_016862441.1 | 8 | 1266 | |||
XM_017006953.1 | XP_016862442.1 | 8 | 1272 | |||
XM_017006954.1 | XP_016862443.1 | 2 | 389 |
Gene Symbol: LOC100132731 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125726.1 | - | 3 | 466 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
