siRNA Details
Gene Symbol: COX19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031617.2 | NP_001026788.1 | 3 | 3137 | ||
GenBank mRNA | AK023185.1 | - | 3 | 3073 | ||
AY957566.1 | - | 3 | 3084 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
