siRNA Details
Gene Symbol: COX19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031617.2 | NP_001026788.1 | 3 | 1581 | ||
GenBank mRNA | AK023185.1 | - | 3 | 1517 | ||
AY957566.1 | - | 3 | 1531 | |||
BC110420.1 | - | 3 | 1518 | 5208612 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
