siRNA Details
Gene Symbol: C2orf74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143959.1 | NP_001137431.1 | 4 | 507 | ||
NM_001143960.2 | NP_001137432.1 | 4 | 520 | |||
NM_001316317.1 | NP_001303246.1 | 3 | 406 | |||
XM_017003967.1 | XP_016859456.1 | 4 | 508 | |||
GenBank mRNA | BP422302.1 | - | 3 | 327 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
