siRNA Details
Gene Symbol: ANKDD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276713.1 | NP_001263642.1 | 9 | 1128 | ||
XM_011543618.2 | XP_011541920.1 | 9 | 1315 | |||
XM_011543623.2 | XP_011541925.1 | 5 | 880 | |||
XM_017009814.1 | XP_016865303.1 | 8 | 1420 | |||
XM_017009815.1 | XP_016865304.1 | 8 | 1420 | |||
XM_017009817.1 | XP_016865306.1 | 4 | 814 | |||
GenBank mRNA | AK307154.1 | - | 3 | 523 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
