siRNA Details
Gene Symbol: ANKDD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276713.1 | NP_001263642.1 | 8, 9 | 1059 | ||
XM_011543618.2 | XP_011541920.1 | 8, 9 | 1246 | |||
XM_011543623.2 | XP_011541925.1 | 4, 5 | 811 | |||
XM_017009814.1 | XP_016865303.1 | 7, 8 | 1351 | |||
XM_017009815.1 | XP_016865304.1 | 7, 8 | 1351 | |||
GenBank mRNA | AK307154.1 | - | 2, 3 | 454 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
