siRNA Details
Gene Symbol: AFDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291964.1 | NP_001278893.1 | 2, 3 | 492 | ||
XM_005266997.2 | XP_005267054.1 | 2, 3 | 555 | |||
XM_006715491.2 | XP_006715554.1 | 2, 3 | 555 | |||
XM_017010878.1 | XP_016866367.1 | 2, 3 | 555 | |||
XM_017010880.1 | XP_016866369.1 | 2, 3 | 539 | |||
XM_017010882.1 | XP_016866371.1 | 2, 3 | 555 | |||
XM_017010883.1 | XP_016866372.1 | 2, 3 | 555 | |||
XM_017010886.1 | XP_016866375.1 | 2, 3 | 555 | |||
GenBank mRNA | AB621809.1 | - | 2, 3 | 492 | ||
AI271860.1 | - | 2, 3 | 441 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
