siRNA Details
Gene Symbol: FAM83H-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091211.1 | - | 1 | 462 | ||
AK125852.1 | - | 1 | 1843 | |||
KF773847.1 | - | 1 | 1552 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
