siRNA Details
Gene Symbol: SPDYE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128076.1 | NP_001121548.1 | 2 | 346 | ||
XM_005256632.4 | XP_005256689.1 | 2 | 1215 | |||
XM_005256633.4 | XP_005256690.1 | 2 | 1215 | |||
GenBank mRNA | BC146949.1 | - | 2 | 346 | 9056877 | |
BC146966.1 | - | 2 | 346 | 9056894 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
