siRNA Details
Gene Symbol: TTC41P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027249.1 | 14 | 3694 | |||
GenBank mRNA | BC064342.1 | - | 4 | 584 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
