siRNA Details
Gene Symbol: DNAH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020877.3 | NP_065928.2 | 37 | 7208 | ||
XM_011523663.1 | XP_011521965.1 | 39 | 6076 | |||
XM_011523664.2 | XP_011521966.1 | 40 | 6225 | |||
XM_011523666.1 | XP_011521968.1 | 39 | 6076 | |||
XM_011523667.2 | XP_011521969.1 | 38 | 5979 | |||
XM_011523668.1 | XP_011521970.1 | 29 | 4459 | |||
XM_011523669.2 | XP_011521971.1 | 27 | 4143 | |||
XM_011523670.2 | XP_011521972.1 | 39 | 6076 | |||
XM_017024218.1 | XP_016879707.1 | 26 | 4160 | |||
XM_017024219.1 | XP_016879708.1 | 39 | 6076 | |||
GenBank mRNA | AB040936.2 | - | 37 | 7208 | ||
BX648163.1 | - | 18 | 2911 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
