siRNA Details
Gene Symbol: KLHL33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109997.2 | NP_001103467.2 | 2 | 961 | ||
XM_005267347.4 | XP_005267404.1 | 3 | 1960 | |||
XM_011536450.2 | XP_011534752.1 | 3 | 1676 | |||
GenBank mRNA | BC146951.1 | - | 1 | 555 | 9056879 | |
BC146954.1 | - | 1 | 555 | 9056882 | ||
BC151130.1 | - | 1 | 577 | 9057016 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
