siRNA Details
Gene Symbol: LDLRAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013693.2 | NP_001013715.2 | 5 | 1410 | ||
GenBank mRNA | AK127275.1 | - | 5 | 1410 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
