siRNA Details
Gene Symbol: TMEM120B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080825.2 | NP_001074294.2 | 2 | 316 | ||
XM_017018853.1 | XP_016874342.1 | 2 | 316 | |||
XM_017018854.1 | XP_016874343.1 | 2 | 316 | |||
GenBank mRNA | AK126568.1 | - | 2 | 316 | ||
BC035884.1 | - | 2 | 133 | |||
BC127768.1 | - | 2 | 194 | 40127741 | ||
BC127769.1 | - | 2 | 194 | 40127743 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
