siRNA Details
Gene Symbol: BSPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128326.2 | NP_001121798.1 | 5 | 465 | ||
XM_017026118.1 | XP_016881607.1 | 5 | 465 | |||
GenBank mRNA | DQ227497.1 | - | 5 | 466 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
