siRNA Details
Gene Symbol: OR7E24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079935.1 | NP_001073404.1 | 1 | 662 | ||
GenBank mRNA | BC146920.1 | - | 1 | 776 | 9056848 | |
Y10529.1 | - | 1 | 437 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
