siRNA Details
Gene Symbol: RNF213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256071.2 | NP_001243000.2 | 26 | 5924 | ||
XM_005257545.4 | XP_005257602.2 | 27 | 6112 | |||
XM_005257546.4 | XP_005257603.2 | 27 | 6016 | |||
XM_006721995.3 | XP_006722058.1 | 27 | 6399 | |||
XM_011525084.2 | XP_011523386.1 | 27 | 6112 | |||
XM_011525086.2 | XP_011523388.1 | 27 | 6112 | |||
XM_011525087.2 | XP_011523389.1 | 27 | 6112 | |||
XM_017024905.1 | XP_016880394.1 | 22 | 4979 | |||
GenBank mRNA | AB537889.1 | - | 26 | 5917 | ||
AK127914.1 | - | 1 | 244 | |||
BX097483.1 | - | 1 | 216 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
