siRNA Details
Gene Symbol: C17orf99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163075.1 | NP_001156547.1 | 3 | 164 | ||
XM_011524152.2 | XP_011522454.1 | 3 | 175 | |||
XM_017023997.1 | XP_016879486.1 | 3 | 178 | |||
XM_017023998.1 | XP_016879487.1 | 3 | 239 | |||
XM_017024000.1 | XP_016879489.1 | 2 | 253 | |||
XM_017024001.1 | XP_016879490.1 | 2 | 523 | |||
GenBank mRNA | AK303128.1 | - | 2 | 82 | ||
AY358510.1 | - | 3 | 171 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
