siRNA Details
Gene Symbol: C17orf99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163075.1 | NP_001156547.1 | 4 | 529 | ||
XM_011524152.2 | XP_011522454.1 | 4 | 540 | |||
XM_017023997.1 | XP_016879486.1 | 4 | 543 | |||
XM_017023998.1 | XP_016879487.1 | 4 | 604 | |||
XM_017023999.1 | XP_016879488.1 | 3 | 310 | |||
XM_017024000.1 | XP_016879489.1 | 3 | 618 | |||
XM_017024001.1 | XP_016879490.1 | 3 | 888 | |||
GenBank mRNA | AK303128.1 | - | 3 | 447 | ||
AY358510.1 | - | 4 | 536 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
