siRNA Details
Gene Symbol: FAM83G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039999.2 | NP_001035088.2 | 5 | 1924 | ||
XM_017024953.1 | XP_016880442.1 | 5 | 2008 | |||
GenBank mRNA | AK123558.1 | - | 3 | 2194 | ||
BX402942.2 | - | 1 | 638 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
