siRNA Details
Gene Symbol: TMEM14EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132656.1 | 1 | 303 | |||
GenBank mRNA | AW866456.1 | - | 1 | 30 | ||
AY358260.1 | - | 1 | 303 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
