siRNA Details
Gene Symbol: TBC1D28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039397.2 | NP_001034486.2 | 10 | 1720 | ||
XM_011523781.2 | XP_011522083.1 | 11 | 1156 | |||
XM_011523782.2 | XP_011522084.1 | 11 | 1036 | |||
XM_017024416.1 | XP_016879905.1 | 10 | 1068 | |||
XM_017024417.1 | XP_016879906.1 | 11 | 1115 | |||
XM_017024418.1 | XP_016879907.1 | 11 | 1213 | |||
XM_017024419.1 | XP_016879908.1 | 10 | 1039 | |||
XM_017024420.1 | XP_016879909.1 | 11 | 2507 | |||
XM_017024421.1 | XP_016879910.1 | 11 | 1086 | |||
XM_017024422.1 | XP_016879911.1 | 10 | 913 | |||
XM_017024423.1 | XP_016879912.1 | 6 | 745 | |||
GenBank mRNA | AK097563.1 | - | 9 | 2008 | ||
AL137747.1 | - | 1 | 547 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
