siRNA Details
Gene Symbol: ZAR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136571.1 | NP_001130043.1 | 3, 4 | 844 | ||
GenBank mRNA | BC146956.1 | - | 3, 4 | 842 | 9056884 | |
BC146965.1 | - | 3, 4 | 844 | 9056893 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
