siRNA Details
Gene Symbol: AP5Z1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014855.2 | NP_055670.1 | 6 | 809 | ||
XM_017012864.1 | XP_016868353.1 | 5 | 636 | |||
GenBank mRNA | AB007875.1 | - | 6 | 788 | ||
BC037399.1 | - | 4 | 482 | 5172434 | ||
CD671081.1 | - | 3 | 233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
