siRNA Details
Gene Symbol: AP5Z1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014855.2 | NP_055670.1 | 16 | 2052 | ||
XM_017012864.1 | XP_016868353.1 | 15 | 1879 | |||
GenBank mRNA | AB007875.1 | - | 16 | 2276 | ||
AK092068.1 | - | 3 | 1515 | |||
BC008841.2 | - | 7 | 720 | |||
BC014240.2 | - | 3 | 243 | |||
BC037399.1 | - | 13 | 2254 | 5172434 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
