siRNA Details
Gene Symbol: C7orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100159.2 | NP_001093629.1 | 9 | 2055 | ||
NM_001267865.1 | NP_001254794.1 | 8 | 1851 | |||
NM_001267866.1 | NP_001254795.1 | 7 | 1839 | |||
XM_006715649.3 | XP_006715712.1 | 7 | 1903 | |||
XM_006715650.2 | XP_006715713.1 | 8 | 1882 | |||
XM_011515121.1 | XP_011513423.1 | 9 | 1959 | |||
XM_011515122.2 | XP_011513424.1 | 8 | 1867 | |||
XM_011515123.2 | XP_011513425.1 | 7 | 1855 | |||
GenBank mRNA | BC031107.1 | - | 9 | 2047 | ||
BC045737.1 | - | 7 | 1839 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
