siRNA Details
Gene Symbol: ZNF717 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128223.2 | NP_001121695.1 | 5 | 1696 | ||
NM_001290208.1 | NP_001277137.1 | 5 | 1677 | |||
NM_001290209.1 | NP_001277138.1 | 5 | 1706 | |||
NM_001324027.1 | NP_001310956.1 | 5 | 1696 | |||
XM_005264711.3 | XP_005264768.1 | 6 | 1774 | |||
XM_006712912.3 | XP_006712975.1 | 5 | 1742 | |||
XM_011533244.2 | XP_011531546.1 | 6 | 1624 | |||
XM_011533245.2 | XP_011531547.1 | 6 | 1760 | |||
XM_011533246.2 | XP_011531548.1 | 6 | 1625 | |||
XM_011533248.2 | XP_011531550.1 | 6 | 1625 | |||
XM_011533249.2 | XP_011531551.1 | 6 | 1623 | |||
GenBank mRNA | AF226994.1 | - | 1 | 301 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
