siRNA Details
Gene Symbol: TDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001578.1 | 4 | 340 | |||
GenBank mRNA | AJ301562.1 | - | 4 | 340 | ||
AK057762.1 | - | 5 | 429 | |||
AY101186.1 | - | 3 | 242 | |||
AY101187.1 | - | 3 | 242 | |||
BC074983.2 | - | 4 | 277 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
