siRNA Details
Gene Symbol: SVIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320340.1 | NP_001307269.1 | 4 | 630 | ||
NM_001320342.1 | NP_001307271.1 | 4 | 514 | |||
NM_148893.2 | NP_683691.1 | 4 | 521 | |||
NR_135213.1 | 4 | 476 | ||||
GenBank mRNA | AF527534.1 | - | 4 | 495 | ||
AL833119.1 | - | 4 | 494 | |||
BG531443.1 | - | 4 | 457 | |||
BI826588.1 | - | 4 | 629 | |||
DA306370.1 | - | 4 | 471 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
