siRNA Details
Gene Symbol: ZNF674-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015378.3 | 2 | 1075 | |||
GenBank mRNA | AK056314.1 | - | 2 | 1046 | ||
BC034616.1 | - | 2 | 1072 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
