siRNA Details
Gene Symbol: FAM78B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017961.4 | NP_001017961.1 | 2 | 1011 | ||
NR_135199.1 | 2 | 1011 | ||||
XM_017000417.1 | XP_016855906.1 | 2 | 1011 | |||
XM_017000418.1 | XP_016855907.1 | 2 | 562 | |||
GenBank mRNA | AK299946.1 | - | 2 | 541 | ||
BC114214.1 | - | 3 | 1094 | 5588830 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
