siRNA Details
Gene Symbol: NCKAP5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037806.3 | NP_001032895.2 | 8 | 2966 | ||
XM_005269050.3 | XP_005269107.1 | 8 | 3291 | |||
XM_006719525.2 | XP_006719588.1 | 8 | 3261 | |||
XM_011538610.2 | XP_011536912.1 | 9 | 3045 | |||
XM_011538611.2 | XP_011536913.1 | 9 | 3015 | |||
GenBank mRNA | AB046822.1 | - | 1 | 1907 | ||
AF370366.1 | - | 1 | 274 | |||
BC007998.1 | - | 2 | 485 | |||
BC033253.1 | - | 1 | 1482 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
