siRNA Details
Gene Symbol: NCKAP5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037806.3 | NP_001032895.2 | 13 | 4169 | ||
XM_006719525.2 | XP_006719588.1 | 13 | 4464 | |||
XM_011538610.2 | XP_011536912.1 | 14 | 4248 | |||
XM_011538611.2 | XP_011536913.1 | 14 | 4218 | |||
GenBank mRNA | AB046822.1 | - | 5 | 3504 | ||
AF370366.1 | - | 6 | 1472 | |||
BC007998.1 | - | 7 | 1688 | |||
BC033253.1 | - | 6 | 2685 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
