siRNA Details
Gene Symbol: LRRIQ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105659.1 | NP_001099129.1 | 7 | 1843 | ||
XM_011540658.1 | XP_011538960.1 | 6 | 1533 | |||
XM_011540659.1 | XP_011538961.1 | 5 | 1047 | |||
XM_011540660.1 | XP_011538962.1 | 5 | 3172 | |||
XM_011540662.2 | XP_011538964.1 | 4 | 1471 | |||
GenBank mRNA | BC062795.1 | - | 1 | 571 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
